3-Methoxypyridin-4-amine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR46502-100MG
OR46502-250MG
OR46502-500MG
76585-036EA
63.16
USD
76585-036
76585-038
76585-040
3-Methoxypyridin-4-amine
3-Methoxypyridin-4-amine
Formula:
C6H8N2O Storage Temperature: Ambient |
MDL Number:
MFCD09991938 CAS Number: 52334-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...