5-Bromo-2-iodobenzoic acid 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR4590-100G OR4590-25G OR4590-5G
76584-786EA 67.26 USD
76584-786 76584-784 76584-782
5-Bromo-2-iodobenzoic acid 97+%
5-Bromo-2-iodobenzoic acid
Formula: C₇H₄BrIO₂
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00079717
CAS Number: 21740-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR