2-Piperazinone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Piperazin-2-one

OR4565-5G
76584-752EA 13.39 USD
76584-752 76584-756 76584-754
2-Piperazinone
2-Piperazinone
Formula: C₄H₈N₂O
MW: 100.12 g/mol
Boiling Pt: 164 °C/5.25mmHg
Melting Pt: 134…140 °C
Storage Temperature: Ambient
MDL Number: MFCD01318687
CAS Number: 5625-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR