2-Bromo-4,5-dimethoxybenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR4517-1G
76584-584EA 20.87 USD
76584-584 76584-588 76584-586
2-Bromo-4,5-dimethoxybenzoic acid
2-Bromo-4,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00017542
CAS Number: 6286-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR