2-Phenoxyethanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether

OR4481-25G
76584-052EA 20.87 USD
76584-052
2-Phenoxyethanol
2-Phenoxyethanol
Formula: C₆H₅OCH₂CH₂OH
MW: 138.17 g/mol
Boiling Pt: 245
Melting Pt: 13
Density: 1.107 g/cm³ (25 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002857
CAS Number: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR