Tetrakis(triphenylphosphine)palladium(0)
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
OR4225-1G
OR4225-25G
OR4225-5G
76583-512EA
27.27
USD
76583-512
76583-516
76583-514
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium (0)
Formula:
C₇₂H₆₀P₄Pd Melting Pt: 103…107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 EINECS: 238-086-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...