3-Bromophenylacetylene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-3-ethynylbenzene

OR42232-1G OR42232-25G OR42232-5G
76583-502EA 20.87 USD
76583-502 76583-506 76583-504
3-Bromophenylacetylene
3-Bromophenylacetylene
Formula: C₈H₅Br
MW: 181.029 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03839983
CAS Number: 766-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR