Kugelmeiers Spherical plate 5D Three-Dimensional Cell Culture Plate, Heidolph
Supplier: Heidolph NA, LLC
036140001
76583-036EA 133.2 USD
76583-036
Kugelmeiers Spherical plate 5D Three-Dimensional Cell Culture Plate, Heidolph
Microplates Cell Culture Plates
The spherical plate 5D is a sterilized 3D cell culture plate that can produce up to 9000 uniform, size-controlled, standardized spheroids.
  • Standard 24 well plate dimension (12 wells have microwells (3D spheroid production), 12 wells do not have microwells (control))
  • Each well has 750 microwells, allowing the plate to produce up to 9000 spheroids
  • Plates are sterilized and have pre-applied anti-adherence coating, making them ready to use out of the box
  • There are only three main steps in the SOP and it just takes 24 hours for your 9000 spheroids to be produced
  • A wide range of cell lines, islet cells, and stem cells have been used on this plate successfully
The patented geometry of the microwell standardizes spheroid production: the spheroids are uniform with other spheroids and the individual spheroids have almost perfect spherical dimensions.
Certifications: Guaranteed sterility assurance level (SAL) 10⁻⁶ according to ISO 11137.