Qlicksmart BladeFlask Blade Remover, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
48QS
76583-020EA 84.24 USD
76583-020
Qlicksmart BladeFlask Blade Remover, Cincinnati Surgical
Blade Removers and Dispensers
BladeFlask remover for size 6 to 26 blades.
  • Holds 100 blades
Remove, contain, and dispose of scalpel blades in one click with the BladeFlask. Secured using a reusable mounting bracket, this safety device can help you quickly and safely remove up to 100 blades at the point of use.