Sol-M® Standard Syringes without Needle
Supplier: SOL-MILLENNIUM MEDICAL
P180003 P180003ST P180005 P180005ST P180010 P180010ST P180011 P180020 P180020ST P180030 P180060 180060CT P180001PP P180011LDSNSB P180020NSB P180020STNSB P180030NSB P180060NSB 180060CTNSB P180001PPNSB P180003NSB P180003STNSB P180005NSB P180005STNSB P180010NSB P180010STNSB P180060ST P180030STNSB P180060STNSB
76582-794BX 12.63 USD
76582-794 76582-796 76582-800 76582-802 76582-806 76582-808 76582-812 76582-818 76582-820 76582-824 76582-828 76582-630 76582-792 76645-680 76645-682 76645-684 76645-686 76645-688 76645-656 76645-666 76645-668 76645-670 76645-672 76645-674 76645-676 76645-678 77021-008 77889-288 77889-290
Sol-M® Standard Syringes without Needle
Syringes Syringes, Medical
3-Piece syringes accurately measure liquid medications and are available in a range of sizes and connection combinations to address a wide variety of clinical needs.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
Available in Luer lock, Luer slip tip and catheter tip.