Sol-Care® Safety Needle with Syringe Combination, Sol M
Supplier: SOL-MILLENNIUM MEDICAL
102115SN 102215SN 12310SN 32510SN 32515SN 32558SN 52115SN 52215SN 32110SN 32115SN 32210SN 32215SN 32310SN
76582-560BX 30.1 USD
76582-560 76582-562 76582-574 76582-734 76582-736 76582-738 76582-740 76582-742 76582-724 76582-726 76582-728 76582-730 76582-732
Sol-Care® Safety Needle with Syringe Combination, Sol M
Syringes Syringes, Medical
This combination is equipped with a pivoting safety shield mechanism that locks in place to lower the risk of accidental injury after medication administration.
  • Shield swings over the needle to cover after use, preventing needlestick injuries
  • Syringe is placed side-by-side within a blister pack for clinician convenience
  • 1 ml syringe feature 0.01 ml graduations
Syringes come equipped with low dead space technology to minimize medication waste. Average 1ml safety needle combination dead space is <0.016 ml.