VWR® General Purpose Laboratory Refrigerator/Freezer Combo Unit with Manual Defrost (30 cu. ft.)
Supplier: VWR International
76580-206
76580-206EA 8745.27 USD
76580-206
VWR® General Purpose Laboratory Refrigerator/Freezer Combo Unit with Manual Defrost (30 cu. ft.)
Refrigerators
These VWR® general purpose laboratory combo units provide everyday features for multi-functional use.
  • Mechanical thermostat
  • Keyed door lock
  • High density foam insulation
  • Three adjustable shelves (refrigerator)
  • Probe access ports for both refrigerator and freezer
These laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with mechanical thermostats, keyed door locks, interior shelving and probe acess. These general purpose refrigerators provide multi-functional features in a cost-effective design.
Material: White powder coat
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.