VWR® Plus Flammable Storage Laboratory Refrigerator Freezer Combo Unit (10 cu. ft.)
Supplier: VWR International
76580-168
76580-168EA 5000.17 USD
76580-168
VWR® Plus Flammable Storage Laboratory Refrigerator Freezer Combo Unit (10 cu. ft.)
Refrigerators
These VWR® laboratory cold storage units are designed for the safe storage of flammable substances.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible and visual alarm systems
  • Remote alarm contacts
  • Keyed door lock
These flammable storage combo units are designed to store and protect volatile substances in laboratory settings. These units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
Material: White powder coat
Certifications: Units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. UL/C-UL listed.
Ordering information: Units come with a two year parts and labor warranty.