VWR® Standard Flammable Storage Laboratory Refrigerator (30 cu. ft.)
Supplier: VWR International
VWR-FRS-30
76580-146EA 7758.63 USD
76580-146
VWR® Standard Flammable Storage Laboratory Refrigerator (30 cu. ft.)
Refrigerators
These VWR® laboratory refrigerators are designed for the safe storage of flammable substances.
  • Mechanical thermostat
  • Keyed door lock
  • High density foam insulation
These flammable storage refrigerators are designed to store and protect volatile substances in laboratory settings. These units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
The standard line provides multi-functional features in a cost-effective design.
Material: White powder coat
Certifications: Units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. UL/C-UL listed.
Ordering information: Units come with a two year parts and labor warranty.