VWR® Plus Flammable Storage Laboratory Refrigerator (30 cu. ft.)
Supplier: VWR International
VWR-FRP-30
76580-142EA 9775.45 USD
76580-142
VWR® Plus Flammable Storage Laboratory Refrigerator (30 cu. ft.)
Refrigerators
These VWR® laboratory refrigerators are designed for the safe storage of flammable substances.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible and visual alarm systems
  • Alarm validation and alarm mute with ring back
  • Keyed door lock
These flammable storage refrigerators are designed to store and protect volatile substances in laboratory settings. These units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
The plus line includes premium additions such as extensive alarm systems and digital touch pad displays.
Material: White powder coat.
Certifications: Units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. UL/C-UL listed.
Ordering information: Units come with a two year parts and labor warranty.