VWR® Standard Flammable Storage Laboratory Freezer (30 cu. ft.)
Supplier: VWR International
76580-138
76580-138EA 8012.55 USD
76580-138
VWR® Standard Flammable Storage Laboratory Freezer (30 cu. ft.)
Freezers Flammable Material Freezers
These VWR® laboratory freezers are designed for the safe storage of flammable substances.
  • Mechanical thermostat
  • Keyed door lock
  • High density foam insulation
These flammable storage freezers are designed to store and protect volatile substances in laboratory settings. These freezers meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
The standard line provides multi-functional features in a cost-effective design.
Material: White powder coat
Certifications: Units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. UL/C-UL listed.
Ordering information: Units come with a two year parts and labor warranty.