VWR® Standard Hazardous Location (Explosion Proof) Laboratory Refrigerator with Manual Defrost (30 cu. ft.)
Supplier: VWR International
76580-132
76580-132EA 8674.67 USD
76580-132
VWR® Standard Hazardous Location (Explosion Proof) Laboratory Refrigerator with Manual Defrost (30 cu. ft.)
Refrigerators
They utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
  • Mechanical thermostat sealed in explosion proof box
  • Keyed door lock
  • High density foam insulation
  • Must be hardwired for safety
These units are equipped with interior shelving, and microprocessor temperature controllers. The standard line provides multi-functional features in a cost-effective design.
Material: White powder coat
Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups C and D guidelines.
Ordering information: Units come with a two year parts and labor warranty.
Caution: Units are not equipped with a line cord and plug. They must be hardwired in conduit to 120 VAC power source for safety.