VWR® General Purpose Laboratory Refrigerator Solid Door (30 cu. ft.)
Supplier: VWR International
VWR-30R
76580-126EA 6599.84 USD
76580-126
VWR® General Purpose Laboratory Refrigerator Solid Door (30 cu. ft.)
Refrigerators
These VWR® general purpose laboratory refrigerators provide everyday features for multi-functional use.
  • Mechanical thermostat
  • Keyed door lock
  • High density foam insulation
  • Five adjustable shelves
  • Probe access port
These laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with mechanical thermostats, keyed door locks, adjustable shelving and probe acess. These general purpose refrigerators provide multi-functional features in a cost-effective design.
Material: White powder coat
Certifications: UL/C-UL listed.