OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
OSMOTECH_PRO
76579-778EA 127467.68 USD
76579-778
OsmoTECH® PRO Multi-Sample Micro-Osmometer, Advanced Instruments
Clinical Diagnostic Supplies Clinical Analysers
Optimize and ensure the quality and consistency of buffers, media, product, excipients, and process parameters to maximize biologic yield, quality, and purity.
  • Multi-sample osmometer
  • Gold standard freezing point technology
  • Fully compliant with 21 CFR part 11, GMP, and EU Annex 11 with e-Signature
  • Meets Pharmacopeia osmolality testing guideline
Designed to meet the high sample throughput, regulatory compliance (including 21 CFR part 11), and data management needs of busy bioprocessing and biomanufacturing labs, the OsmoTECH PRO Multi-Sample Micro-Osmometer combines ease-of-use with accurate and precise osmolality measurements for reliable and reproducible results. Optimize and ensure the quality and consistency of buffers, media, product, excipients, and process parameters to maximize biologic yield, quality and purity.
Accessories information: OsmoTECH® PRO Multi-Sample Micro-Osmometer IQ/OQ Service (sold separately): Eliminate delays with our IQ/OQ. Our comprehensive services ensure no workflow interruption so you can focus on your work. Our experts developed this comprehensive qualification package to comply with USP <1058> Analytical Instrument Qualification and USP <785> Osmolality and Osmolarity Testing. Don’t go it alone. Regulated environments require documented proof that your manufacturing process consistently produces quality product. Performing qualification internally is costly, time-consuming, and problematic. Our expert qualification services ensure fast and reliable commissioning of your instrument. Eliminate delays with our IQ/OQ. Our comprehensive services ensure no workflow interruption so you can focus on your work.