Edge Utility Cutter, Slice®
Supplier: SLICE, INC.
10479
76579-610BX 403.25 USD
76579-610
Edge Utility Cutter, Slice®
Knives
Edge utility cutter is well crafted and sturdy. Excellent for any cut that requires a clamping motion, like snipping wires, the tool will impress you with its smooth and steady movement. The comfortable soft-touch handle includes a Blade Garage™ for extra capacity and a lock for safe tool storage.
  • Ambidextrous design
  • Chemically inert blade never rusts
  • Oil and lubricant free blade
  • 100% zirconium oxide blade
  • Blade edge lasts up to 11× longer than metal
  • Fewer blade changes - fewer injuries
  • No-tool blade change
This edge utility cutter is incredibly well crafted and sturdy. It is compatible with two different styles of Slice safety blade: the Utility Knife blades (10526 and 10528) and the Utility Blades (10523, 10524, 10525). Excellent for any cut that requires a clamping motion, like snipping wires, the edge utility cutter by Slice will impress you with its smooth and steady movement. The comfortable soft-touch handle includes a Blade Garage™ for extra capacity and a lock for safe tool storage.