Small Pathology Containers, Mopec
Supplier: Mortech Manufacturing
BG640 BG660 BG690 BG610 BG620
76579-436CS 559.94 USD
76579-436 76579-442 76579-450 76579-416 76579-426
Small Pathology Containers, Mopec
Sample Containers General Purpose Containers
Mopec’s pathology containers are manufactured from high-quality, clarified polypropylene and feature a closure that guarantees the container’s seal is fully secured. Containers come empty and are available in sizes 20 - 120 ml.
  • Polypropylene container, with polyethylene screw cap
  • Features a closure that guarantees the container’s seal is fully secured
Sold by the case.