ALCH reagent 100%

Supplier: MORTECH MANUFACTURING
Danger

Synonyms: Reagent alcohol

BG736
76579-360CS 494.58 USD
76579-360
ALCH reagent 100%
Alcohol

1 gallon container of reagent alcohol, 100%

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR