ALCH reagent 80%

Supplier: MORTECH MANUFACTURING
Danger

Synonyms: Reagent alcohol

BG734
76579-356CS 412.13 USD
76579-356
ALCH reagent 80%
Alcohol

1 gallon container of reagent alcohol, 80%

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR