Alcohol reagent 50%

Supplier: Mopec
Danger

BG732
76579-352CS 354.61 USD
76579-352
Alcohol reagent 50%
Alcohol

1 gallon container of reagent alcohol, 50%

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR