Formalin prefilled 10%

Supplier: MORTECH MANUFACTURING
Warning

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

BG515 BG516 BG518 BG524 BG548 BG500 BG502 BG504 BG505 BG506 BG507 BG509 BG512
76579-330EA 65.26 USD
76579-330 76579-332 76579-334 76579-338 76579-344 76579-312 76579-316 76579-318 76579-320 76579-322 76579-324 76579-326 76579-328
Formalin prefilled 10%
Formalin

Mopec’s Prefilled Formalin Containers are specifically designed to safely collect, store and transport various sizes of tissue specimens. Containers are half-filled with a 10% neutral buffered formalin fixative and come in a variety of sizes.


  • Filled with a 10% neutral buffered formalin
  • General purpose fixative for tissues.
  • Penetrates quickly, but fixes slowly
  • No other dilution or additional agents are necessary

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR