Mini Slide Drying Hotplate, Mopec
Supplier: Mortech Manufacturing
BK402
76579-284EA 1197.68 USD
76579-284
Mini Slide Drying Hotplate, Mopec
Slide Warmers/Dryers
Mopec’s mini slide drying hotplate is designed to quickly and efficiently remove wrinkles from tissue sections while taking up minimal bench-top space. The rapid heating system ensures the hotplate is ready to use in less time.
  • Designed to take up minimal bench-top space
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
Sold individually.