Heated Tampers, Mopec
Supplier: Mopec
BK351 BK352 BK353
76579-270EA 591.02 USD
76579-270 76579-272 76579-274
Heated Tampers, Mopec
Presses
Mopec’s heated tampers are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.
  • Molded finger grips increase user comfort and remain cool when in use
  • Bayonet connection prevents disconnection while in use
  • Compatible with the Mopec control unit (BK330)
Available in three sizes and compatible with the Mopec control unit.