Heated Forceps - LED, Mopec
Supplier: Mortech Manufacturing
BK301L-SM BK302L-SE BK302L-SM BK301L-SE BK304L-SE BK304L-SM
76579-256EA 766.81 USD
76579-256 76579-252 76579-258 76579-250 76579-254 76579-260
Heated Forceps - LED, Mopec
Forceps
Mopec’s heated forceps with LED are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.
  • LED Light to assist with viewing
  • Designed to take up minimal bench-top space
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Molded finger grips increase user comfort and remain cool when in use
  • Bayonet connection prevents disconnection while in use
The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth. Choose between serrated or smooth tips. Available in three sizes and compatible with the Mopec control unit.