Round Tissue Flotation Bath
Supplier: Mortech Manufacturing
BK100
76579-210EA 1485.44 USD
76579-210
Round Tissue Flotation Bath
Baths Tissue Baths
Mopec’s Round Tissue Flotation Bath is designed with a deep center bowl so slides can be fully submerged at a convenient angle. It features an anti-microbial coating to protect the system from bacterial and viral growth as well as a rapid heating system to reduce waiting time.
  • Designed to take up minimal bench-top space
  • Deep bowl to assist with tissue pick-up at any angle
  • Rapid heating system ensures system is ready to use in less time
  • Analog temperature control
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps improve temperature control and prevent surface particles (76579-212)