3-Amino-2-bromo-4-methylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR30941-1G
76575-366EA 20.32 USD
76575-366 76575-368
3-Amino-2-bromo-4-methylpyridine 97%
3-Amino-2-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD04112497
CAS Number: 126325-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR