2-Chloro-4,6-dimethylpyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR30935-5G OR30935-25G OR30935-100G
76575-336EA 23.44 USD
76575-336 76575-338 76575-340
2-Chloro-4,6-dimethylpyridine 98%
2-Chloro-4,6-dimethylpyridine
Formula: C₇H₈ClN
Storage Temperature: Ambient
MDL Number: MFCD08277279
CAS Number: 30838-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR