1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR307301-5G OR307301-1G
76574-974EA 1440 USD
76574-974 76574-972
1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
Formula: C₇H₁₁IN₂O
Storage Temperature: Ambient
MDL Number: MFCD30721063
CAS Number: 1639858-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR