Bis(triphenylphosphine)palladium(II) chloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Bis(triphenylphosphine)palladium(II) dichloride

OR30692-25G OR30692-1G OR30692-5G
76574-702EA 552.12 USD
76574-702 76574-698 76574-700
Bis(triphenylphosphine)palladium(II) chloride 98%
Bis(triphenylphosphine)palladium (II) chloride
Formula: C₃₆H₃₀Cl₂P₂Pd
Melting Pt: 310 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00009593
CAS Number: 13965-03-2
EINECS: 237-744-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR