(S)-(+)-Ethyl-3-hydroxybutyrate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR304080-1G
76574-228EA 174.07 USD
76574-228 76574-226
(S)-(+)-Ethyl-3-hydroxybutyrate 95%
(S)-(+)-Ethyl 3-hydroxybutyrate
Formula: C₆H₁₂O₃
Boiling Pt: 180…182 °C
Density: 1.012 g/cm³ (25 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00066206
CAS Number: 56816-01-4
EINECS: 260-393-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR