2-Amino-4-methylbenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-p-toluic acid, 4-Methylanthranilic acid, 4-Methylanthranil acid, 2-Amino-4-methylbenzoic Acid

OR30203-5G
76573-920EA 79.31 USD
76573-920 76573-916 76573-918
2-Amino-4-methylbenzoic acid
2-Amino-4-methylbenzoic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00047853
CAS Number: 2305-36-4
EINECS: 218-976-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR