3,4-Dimethylbenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR30113-25G OR30113-100G
76573-752EA 20.26 USD
76573-752 76573-754
3,4-Dimethylbenzaldehyde
3,4-Dimethylbenzaldehyde
Formula: C₉H₁₀O
Boiling Pt: 102…104 °C/15mmHg
Melting Pt: 223…226 °C
Density: 1.012 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016612
CAS Number: 5973-71-7
EINECS: 227-770-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR