2-Amino-5-bromo-3-methylpyrazine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Bromo-3-methylpyrazin-2-amine

OR300250-1G OR300250-250MG
76573-400EA 132.42 USD
76573-400 76573-398
2-Amino-5-bromo-3-methylpyrazine
2-Amino-5-bromo-3-methylpyrazine
Formula: C₅H₆BrN₃
Storage Temperature: Ambient
MDL Number: MFCD08705773
CAS Number: 74290-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR