(3-Bromo-5-methylphenyl)methanamine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR300246-1G OR300246-250MG
76573-384EA 363.66 USD
76573-384 76573-382
(3-Bromo-5-methylphenyl)methanamine hydrochloride
(3-Bromo-5-methylphenyl)methanamine hydrochloride
Formula: C₈H₁₀BrN·ClH
Storage Temperature: Ambient
MDL Number: MFCD28893555
CAS Number: 1646570-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR