5-(2-Pyridyl)-1,3-oxazole

Supplier: APOLLO SCIENTIFIC
Warning

OR29044-1G OR29044-250MG
76572-808EA 166.04 USD
76572-808 76572-806
5-(2-Pyridyl)-1,3-oxazole
5-(2-Pyridyl)-1,3-oxazole
Formula: C₈H₆N₂O
Boiling Pt: 95…98 °C/0.2mmHg
Storage Temperature: Ambient
MDL Number: MFCD00085160
CAS Number: 70380-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR