Dimethyl-3-oxoglutarate

Supplier: APOLLO SCIENTIFIC

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

OR28676-25G
76572-566EA 20.79 USD
76572-566
Dimethyl-3-oxoglutarate
Dimethyl 3-oxoglutarate
Formula: C₇H₁₀O₅
MDL Number: MFCD00008462
CAS Number: 1830-54-2
EINECS: 217-385-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR