2-Morpholino-5-nitrobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR27640-500MG OR27640-100MG
76572-238EA 83.46 USD
76572-238 76572-236
2-Morpholino-5-nitrobenzaldehyde
2-Morpholino-5-nitrobenzaldehyde
Formula: C₁₁H₁₂N₂O₄
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00439921
CAS Number: 30742-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR