Tris(2-methylphenyl)phosphine 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

OR25778-100G OR25778-500G OR25778-25G
76571-694EA 49.92 USD
76571-694 76571-696 76571-692
Tris(2-methylphenyl)phosphine 96%
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
Melting Pt: 123…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR