4-Phenylbutyric acid 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Phenylbutanoic acid
OR2390-500G
OR2390-100G
OR2390-25G
76571-194EA
309.68
USD
76571-194
76571-192
76571-190
4-Phenylbutyric acid 98%
4-Phenylbutyric acid
Formula:
C₁₀H₁₂O₂ Boiling Pt: 165 °C/10mmHg Melting Pt: 49…53 °C Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004403 CAS Number: 1821-12-1 EINECS: 217-341-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...