(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2317-5G
76570-966EA 20.32 USD
76570-966
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formula: C₉H₁₁NO
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00216656
CAS Number: 136030-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR