Diphenylcyclopropenone
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
OR22091-5G
OR22091-1G
76570-522EA
78.44
USD
76570-522
76570-520
Diphenylcyclopropenone
Diphenylcyclopropenone
Formula:
C₁₅H₁₀O MW: 206.24 g/mol Boiling Pt: 407 Melting Pt: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001311 CAS Number: 886-38-4 EINECS: 212-948-4 Merck Index: 12,03366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...