4'-Butylacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

OR21897-25G OR21897-5G OR21897-100G
76570-428EA 54.34 USD
76570-428 76570-426 76570-430
4'-Butylacetophenone
4'-Butylacetophenone
Formula: C₁₂H₁₆O
Boiling Pt: 101…102 °C/1.5mmHg
Density: 0.957 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017500
CAS Number: 37920-25-5
EINECS: 253-715-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR