5-Bromo-2-methyl-4-nitro-1H-imidazole

Supplier: APOLLO SCIENTIFIC
Warning

OR21890-1G
76570-424EA 20.26 USD
76570-424
5-Bromo-2-methyl-4-nitro-1H-imidazole
5-Bromo-2-methyl-4-nitro-1H-imidazole
Formula: C₄H₄BrN₃O₂
Melting Pt: 256…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00156130
CAS Number: 18874-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR