1-Bromo-3,5-dichlorobenzene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Dichlorobromobenzene

OR19527-25G OR19527-100G OR19527-250G
76569-810EA 20.77 USD
76569-810 76569-812 76569-814
1-Bromo-3,5-dichlorobenzene
1-Bromo-3,5-dichlorobenzene
Formula: C₆H₃BrCl₂
Boiling Pt: 232 °C
Melting Pt: 73…77 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR