Tetraphenylethylene

Supplier: APOLLO SCIENTIFIC
Warning

OR18539-5G
76569-480EA 35.66 USD
76569-480
Tetraphenylethylene
Tetraphenylethylene
Formula: C₂₆H₂₀
Boiling Pt: 420 °C
Melting Pt: 222…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00004764
CAS Number: 632-51-9
EINECS: 211-179-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR