Guaifenesin 99+%

Supplier: APOLLO SCIENTIFIC
Warning

OR18315-1KG
76569-188EA 99.82 USD
76569-188
Guaifenesin 99+%
Guaifenesin
Formula: C₁₀H₁₄O₄
Boiling Pt: 215 °C/15mmHg
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00016873
CAS Number: 93-14-1
EINECS: 202-222-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR